Skip to Content
Merck
All Photos(1)

Key Documents

SML1150

Sigma-Aldrich

SNX-482 Recombinant

≥98% (HPLC)

Synonym(s):

GVDKAGCRYMFGGCSVNDDCCPRLGCHSLFSYCAWDLTFSD

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
€233.00

€233.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 μG
€233.00

About This Item

Empirical Formula (Hill Notation):
C192H274N52O60S7
CAS Number:
Molecular Weight:
4495.00
UNSPSC Code:
12352200
NACRES:
NA.77

€233.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

(from Hysterocrates gigas (African tarantula))

Quality Level

recombinant

expressed in E. coli

Assay

≥98% (HPLC)

form

lyophilized powder

solubility

aqueous buffer: soluble

storage temp.

−20°C

SMILES string

S1SCC2NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C3N(CCC3)C(=O)C4NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(CSSC4)NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CN)C(C)C)CC(=O)O)CCCCN)C)CCCN\C(=N\[H])\N)C

InChI

1S/C192H274N52O60S7/c1-91(2)57-114-157(270)207-79-146(257)213-135-85-306-307-86-136-183(296)237-133(83-248)179(292)242-153(95(9)10)187(300)231-124(67-141(195)252)170(283)228-126(69-148(260)261)173(286)229-127(70-149(262)263)174(287)239-138(184(297)240-139(189(302)244-55-30-42-140(244)185(298)217-112(161(274)219-114)41-29-54-202-192(198)199)89-311-308-84-134(212-145(256)78-205-155(268)96(11)209-159(272)110(39-26-27-52-193)214-171(284)128(71-150(264)265)232-186(299)152(94(7)8)241-142(253)73-194)181(294)215-111(40-28-53-201-191(196)197)160(273)223-119(63-102-43-47-106(250)48-44-102)164(277)216-113(51-56-305-14)162(275)222-117(60-99-31-18-15-19-32-99)158(271)206-76-143(254)204-77-144(255)211-136)88-310-309-87-137(238-167(280)120(64-103-45-49-107(251)50-46-103)225-177(290)131(81-246)234-165(278)118(61-100-33-20-16-21-34-100)224-163(276)115(58-92(3)4)221-176(289)130(80-245)236-169(282)123(226-182(135)295)66-105-75-200-90-208-105)180(293)210-97(12)156(269)218-122(65-104-74-203-109-38-25-24-37-108(104)109)168(281)227-125(68-147(258)259)172(285)220-116(59-93(5)6)175(288)243-154(98(13)249)188(301)230-121(62-101-35-22-17-23-36-101)166(279)235-132(82-247)178(291)233-129(190(303)304)72-151(266)267/h15-25,31-38,43-50,74-75,90-98,110-140,152-154,203,245-251H,26-30,39-42,51-73,76-89,193-194H2,1-14H3,(H2,195,252)(H,200,208)(H,204,254)(H,205,268)(H,206,271)(H,207,270)(H,209,272)(H,210,293)(H,211,255)(H,212,256)(H,213,257)(H,214,284)(H,215,294)(H,216,277)(H,217,298)(H,218,269)(H,219,274)(H,220,285)(H,221,289)(H,222,275)(H,223,273)(H,224,276)(H,225,290)(H,226,295)(H,227,281)(H,228,283)(H,229,286)(H,230,301)(H,231,300)(H,232,299)(H,233,291)(H,234,278)(H,235,279)(H,236,282)(H,237,296)(H,238,280)(H,239,287)(H,240,297)(H,241,253)(H,242,292)(H,243,288)(H,258,259)(H,260,261)(H,262,263)(H,264,265)(H,266,267)(H,303,304)(H4,196,197,201)(H4,198,199,202)

InChI key

NSUPRLHDCFNOKD-UHFFFAOYSA-N

General description

Disulfide bonds between Cys7-Cys21, Cys14-Cys26, and Cys20-Cys33.

Biochem/physiol Actions

SNX-482 is a peptidyl toxin originally isolated from the venom of the spider Hysterocrates gigas. SNX-482 specifically blocks Cav2.3 (a1E, R-type) calcium channels in a voltage dependent manner. SNX-482 also blocks P/Q-type voltage-gated Ca2+ channels and blocks A-type K+ channels (IC50 of <3 nm).
SNX-482 specifically blocks Cav2.3 calcium channels.

Reconstitution

Any aqueous buffer. Centrifuge before use (10000 x g for 5 min). After reconstitution, store for 3 months at −20 °C or up to four weeks at 2-8 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service