Skip to Content
Merck
All Photos(1)

Documents

SML1140

Sigma-Aldrich

ML138

≥98% (HPLC)

Synonym(s):

2-[5-[[(3,4-Dichlorophenyl)methyl]thio]-4-(2-furanylmethyl)-4H-1,2,4-triazol-3-yl]-pyridine, CID 44601470, CID44601470, ML-138

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14Cl2N4OS
CAS Number:
Molecular Weight:
417.31
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC(CSC2=NN=C(C3=NC=CC=C3)N2CC4=CC=CO4)=CC=C1Cl

InChI

1S/C19H14Cl2N4OS/c20-15-7-6-13(10-16(15)21)12-27-19-24-23-18(17-5-1-2-8-22-17)25(19)11-14-4-3-9-26-14/h1-10H,11-12H2

InChI key

QDXGAJIGONMOCL-UHFFFAOYSA-N

Biochem/physiol Actions

ML138 (CID 44601470) is a highly selective agonist at kappa opioid receptors (KOR) that potently activate G protein coupling but weakly interact with β arrestin2.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service