Skip to Content
Merck
All Photos(1)

Key Documents

SML1126

Sigma-Aldrich

PDP-EA

≥98% (HPLC)

Synonym(s):

3-n-Pentadecylphenolethanolamide, N-(2-Hydroxyethyl)-2-(3-pentadecylphenoxy)-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H43NO3
CAS Number:
Molecular Weight:
405.61
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

OCCNC(COC1=CC(CCCCCCCCCCCCCCC)=CC=C1)=O

InChI

1S/C25H43NO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-23-17-15-18-24(21-23)29-22-25(28)26-19-20-27/h15,17-18,21,27H,2-14,16,19-20,22H2,1H3,(H,26,28)

InChI key

BIOVSNWTCKRADD-UHFFFAOYSA-N

Biochem/physiol Actions

3-N-Pentadecylphenolethanolamide (PDP-EA) is an activator of fatty acid amide hydrolase (FAAH) from plant and mammalian species. PDP-EA appears to enhance FAAH activity by reduction of negative feedback from free ethanoloamine.
PDP-EA is an activator of fatty acid amide hydrolase (FAAH).

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service