Skip to Content
Merck
All Photos(1)

Key Documents

SML1096

Sigma-Aldrich

TH588 hydrochloride

≥98% (HPLC)

Synonym(s):

N4-Cyclopropyl-6-(2,3-dichlorophenyl)pyrimidine-2,4-diamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€157.00
25 MG
€598.00

€157.00


Check Cart for Availability


Select a Size

Change View
5 MG
€157.00
25 MG
€598.00

About This Item

Empirical Formula (Hill Notation):
C13H12Cl2N4 · HCl
CAS Number:
Molecular Weight:
331.63
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€157.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

ClC1=CC=CC(C2=NC(N)=NC(NC3CC3)=C2)=C1Cl.Cl

InChI

1S/C13H12Cl2N4.ClH/c14-9-3-1-2-8(12(9)15)10-6-11(17-7-4-5-7)19-13(16)18-10;/h1-3,6-7H,4-5H2,(H3,16,17,18,19);1H

InChI key

FBHMEHNWFXCSKE-UHFFFAOYSA-N

Biochem/physiol Actions

TH588 is a potent inhibitor of human 7,8-Dihydro-8-oxoguaninetriphosphatase MTH1 (NUDT1) with an IC50 value of 5 nM and good metabolic stability. MTH1 hydrolyzes oxidized purine nucleoside triphosphates that might otherwise be incorporated into DNA/RNA and contribute to DNA damage. MTH1 removal of oxidized nucleotides that result from increased levels of reactive oxygen species (ROS) in fast-proliferating cancer cells helps protect cancer cells from proliferative stress and prevent cancer cell death. TH588 is considered a new target for cancer therapy. TH588 is highly selective towards MTH1, with no relevant inhibition of other members of the nudix protein family or a panel of 87 enzymes, GPCRs, kinases, ion channels and transporter. TH588 has been shown to selectively kill a variety of cancer cell lines and with in vivo activity shown for TH588 in SW480 colorectal and MCF7 breast tumour xenografts.
TH588 is a potent inhibitor of human 7,8-Dihydro-8-oxoguaninetriphosphatase MTH1 (NUDT1).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Govindi J Samaranayake et al.
Molecular cancer therapeutics, 19(2), 432-446 (2019-11-21)
Investigations into the human 8-oxodGTPase, MutT Homolog 1 (MTH1), have risen sharply since the first-in-class MTH1 inhibitors were reported to be highly tumoricidal. However, MTH1 as a cancer therapeutic target is currently controversial because subsequently developed inhibitors did not exhibit

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service