Skip to Content
Merck
All Photos(1)

Key Documents

SML1061

Sigma-Aldrich

PF-04856264

≥98% (HPLC)

Synonym(s):

3-Cyano-4-[2-(1-methyl-1H-pyrazol-5-yl)phenoxy]-N-2-thiazolyl-benzenesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€149.00
25 MG
€341.00

€149.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€149.00
25 MG
€341.00

About This Item

Empirical Formula (Hill Notation):
C20H15N5O3S2
CAS Number:
Molecular Weight:
437.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€149.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(C(C=C1)=CC(C#N)=C1OC2=CC=CC=C2C3=CC=NN3C)(NC4=NC=CS4)=O

InChI

1S/C20H15N5O3S2/c1-25-17(8-9-23-25)16-4-2-3-5-19(16)28-18-7-6-15(12-14(18)13-21)30(26,27)24-20-22-10-11-29-20/h2-12H,1H3,(H,22,24)

InChI key

MKSKJVIBSRUWSZ-UHFFFAOYSA-N

Biochem/physiol Actions

PF-04856264 is a potent, selective inhibitor of the human Nav1.7 voltage gated sodium channel (IC50 = 28 nM). PF-04856264 blocks mouse Nav1.7 (IC50 = 131 nM) but has low potency against the rat Nav1.7 channel (IC50 = 4.2 mM).
PF-04856264 is a potent, selective inhibitor of the human Nav1.7 voltage gated sodium channel.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service