Skip to Content
Merck
All Photos(1)

Key Documents

SML1025

Sigma-Aldrich

Lorcainide hydrochloride

≥98% (HPLC)

Synonym(s):

Lorcamide hydrochloride, N-(4-Chlorophenyl)-N-[1-(1-methylethyl)-4-piperidinyl]-benzeneacetamide hydrochloride, R15889 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€87.60
50 MG
€239.00

€87.60


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€87.60
50 MG
€239.00

About This Item

Empirical Formula (Hill Notation):
C22H27ClN2O · HCl
CAS Number:
Molecular Weight:
407.38
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

€87.60


Estimated to ship on24 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)N(C3CCN(CC3)C(C)C)C(=O)Cc2ccccc2.Cl

InChI

1S/C22H27ClN2O.ClH/c1-17(2)24-14-12-21(13-15-24)25(20-10-8-19(23)9-11-20)22(26)16-18-6-4-3-5-7-18;/h3-11,17,21H,12-16H2,1-2H3;1H

InChI key

FSFSWNBDCJVFGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lorcainide is a Class 1c antiarrhythmic agent that blocks open fast acting voltage-gated sodium channels (subtype Nav1.5).
Lorcainide is a Class 1c antiarrhythmic agent that blocks open fast acting voltage-gated sodium channels (subtype Nav1.5). Data shows that Lorcainide is a potent, selective antagonists/inverse agonists of human and rat H3 histamine receptors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service