Skip to Content
Merck
All Photos(1)

Key Documents

SML0999

Sigma-Aldrich

ME0328

≥98% (HPLC)

Synonym(s):

(S)-3,4-Dihydro-4-oxo-N-(1-phenylethyl)-2-quinazolinepropanamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€96.10

€96.10


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€96.10

About This Item

Empirical Formula (Hill Notation):
C19H19N3O2
CAS Number:
Molecular Weight:
321.37
UNSPSC Code:
12352200
NACRES:
NA.77

€96.10


Estimated to ship on20 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

[nH]1c2c([c](nc1CCC(=O)N[C@@H](C)c3ccccc3)=O)cccc2

InChI

1S/C19H19N3O2/c1-13(14-7-3-2-4-8-14)20-18(23)12-11-17-21-16-10-6-5-9-15(16)19(24)22-17/h2-10,13H,11-12H2,1H3,(H,20,23)(H,21,22,24)/t13-/m0/s1

InChI key

QIHBWVVVRYYYRO-ZDUSSCGKSA-N

Biochem/physiol Actions

ME0328 is a selective inhibitor of ADP-ribosyltransferase-3/poly(ADP-ribose) polymerase-3 (ARTD3/PARP3), a PARP family member involved in DNA repair.
ME0328 is a selective inhibitor of ADP-ribosyltransferase-3/poly(ADP-ribose) polymerase-3 (ARTD3/PARP3), a PARP family member involved in DNA repair. ME0328 is the most potent isomer of the four ARTD3 ADP-ribosyltransferase inhibitors. ME0328 selectively inhibits ARTD3 (IC50 of 890 nM) with weaker activity against ARTD1 and ARTD2 (IC50 6.3 and 10.8 μM, respectively), and no significant activity (IC50 >30 μM) against any other ARTD enzyme. ME0328 appears to be the first selective inhibitor of ARTD3/PARP3.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service