Skip to Content
Merck

Skip To

SML0963

(E)-3,4,5,4′-Tetramethoxystilbene

≥98% (HPLC)

Synonym(s):

1,2,3-Trimethoxy-5-[(1E)-2-(4-methoxyphenyl)ethenyl]-benzene, DMU 212, MR-4

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C18H20O4
CAS Number:
Molecular Weight:
300.35
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O(C)c1c(cc(cc1OC)\C=C\c2ccc(cc2)OC)OC

InChI

1S/C18H20O4/c1-19-15-9-7-13(8-10-15)5-6-14-11-16(20-2)18(22-4)17(12-14)21-3/h5-12H,1-4H3/b6-5+

InChI key

GGFQQRXTLIJXNY-AATRIKPKSA-N

Biochem/physiol Actions

(E)-3,4,5,4′-Tetramethoxystilbene (DMU 212) is a derivative of resveratrol with anti-angiogenic and anti-tumor properties.
(E)-3,4,5,4′-Tetramethoxystilbene is a derivative of resveratrol; anti-angiogenic; anti-tumor properties.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters