Skip to Content
Merck

Skip To

SML0959

Bis-1,4-(4-methoxybenzenesulfonamidyl)naphthalene

≥98% (HPLC)

Synonym(s):

Compound 16, N,N′-(Naphthalene-1,4-diyl)bis(4-methoxybenzenesulfonamide)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 118,00

About This Item

Empirical Formula (Hill Notation):
C24H22N2O6S2
CAS Number:
Molecular Weight:
498.57
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder

€ 118,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

, white to light yellow to light gray

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Bis-1,4-(4-methoxybenzenesulfonamidyl)naphthalene is a non-covalent inhibitor of the interaction between Kelch-like ECH-associated protein 1 (Keap1) and nuclear factor erythroid 2-related factor 2 (Nrf2).
Bis-1,4-(4-methoxybenzenesulfonamidyl)naphthalene is a non-covalent inhibitor of the interaction between Kelch-like ECH-associated protein 1 (Keap1) and nuclear factor erythroid 2-related factor 2 (Nrf2). The Keap1–Nrf2 protein–protein interaction is considered a critical point of the Keap1–Nrf2–ARE (antioxidant response elements) system that protects cells from oxidative stress. Bis-1,4-(4-methoxybenzenesulfonamidyl)naphthalene prevents Keap1 repression of Nrf2 activation, allowing Nrf2 to initiate antioxidant response element (ARE) to protect the cell. Bis-1,4-(4-methoxybenzenesulfonamidyl)naphthalene has an IC50 of 2.7 micromolar and specifically binds to the Keap1 Kelch-DC domain.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1922SML1152SML1949
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

color

, white to light yellow to light gray

color

white to beige

color

white to beige

color

white to beige


wgk

WGK 3

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters