Skip to Content
Merck

Skip To

SML0945

Nilvadipine

≥98% (HPLC)

Synonym(s):

2-Cyano-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 3-methyl 5-(1-methylethyl) ester, 5-Isopropyl-3-methyl-2-cyano-1,4-dihydro-6-methyl-4-(m-nitrophenyl)-3,5-pyridinedicarboxylate, FK-235, FR-34235, Isopropyl 6-cyano-5-methoxycarbonyl-2-methyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3-carboxylate, Nivadipine, Nivaldipine, SKF-102362

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 49,30
50 mg
Check Cart for Availability
€ 145,00

About This Item

Empirical Formula (Hill Notation):
C19H19N3O6
CAS Number:
Molecular Weight:
385.37
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

€ 49,30


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

room temp

SMILES string

[N+](=O)([O-])c1cc(ccc1)C2C(=C(NC(=C2C(=O)OC)C#N)C)C(=O)OC(C)C

InChI

1S/C19H19N3O6/c1-10(2)28-19(24)15-11(3)21-14(9-20)17(18(23)27-4)16(15)12-6-5-7-13(8-12)22(25)26/h5-8,10,16,21H,1-4H3

InChI key

FAIIFDPAEUKBEP-UHFFFAOYSA-N

General description

Nilvadipine is a dihydropyridine drug.

Biochem/physiol Actions

Nilvadipine is a potent calcium channel blocker that is used for treatment of hypertension. Nilvadipine, as other dihydropyridine calcium antagonists, decreases intracellular calcium concentration causing dilation of the coronary and systemic arteries. Nilvadipine acutely reduced brain Aβ levels in a transgenic mouse model of Alzheimer′s disease (AD).
Nilvadipine is a potent calcium channel blocker; antihypertensive; antianginal.
Nilvadipine, apart from its antihypertensive effect, is known to decrease the accumulation of β-amyloid protein in the brain. Therefore, it is effective in the treatment of Alzheimer′s disease. Nilvadipine also promotes cognitive function in hypertensive patients.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
N0165N149SML1372
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥95% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

storage temp.

room temp

storage temp.

room temp

storage temp.

-

storage temp.

2-8°C

solubility

DMSO: 15 mg/mL, clear

solubility

DMSO: >10 mg/mL

solubility

methanol: 62.5 mg/mL, 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: insoluble, H2O: insoluble

solubility

DMSO: 10 mg/mL, clear

color

white to beige

color

yellow

color

off-white to yellow

color

white to beige


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters