Skip to Content
Merck

SML0928

AD-01 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

AD01 trifluoroacetate salt, NH2-QIRQQPRDPPTETLELEVSPDPAS-OH trifluoroacetate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C115H187N33O42 · xC2HF3O2
CAS Number:
Molecular Weight:
2703.91 (free base basis)
NACRES:
NA.77
UNSPSC Code:
12352202
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

AD-01 is a potent antiangiogenic peptide a FKBPL fragment, which interact with cell-surface receptor CD44. AD-01 inhibits the self-renewal capacity of CD44-positive breast cancer stem cells (BCSC).
AD-01 is an inhibitor of angiogenesis in vitro and in vivo that mediates its activity through CD44.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library