Skip to Content
Merck
All Photos(1)

Key Documents

SML0918

Sigma-Aldrich

CID44216842

≥98% (HPLC)

Synonym(s):

4-[3-(4-Bromophenyl)-5-(3-methoxyphenyl)-3,4-dihydropyrazol-2-yl]benzenesulfonamide, KSC-221-15, KUC103479N-02

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€282.00

€282.00


Estimated to ship on15 April 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€282.00

About This Item

Empirical Formula (Hill Notation):
C22H20BrN3O3S
CAS Number:
Molecular Weight:
486.38
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

€282.00


Estimated to ship on15 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(C1=CC=C(N2C(C3=CC=C(Br)C=C3)CC(C4=CC(OC)=CC=C4)=N2)C=C1)(N)=O

InChI

1S/C22H20BrN3O3S/c1-29-19-4-2-3-16(13-19)21-14-22(15-5-7-17(23)8-6-15)26(25-21)18-9-11-20(12-10-18)30(24,27)28/h2-13,22H,14H2,1H3,(H2,24,27,28)

InChI key

LPUYDLXQQMWRLR-UHFFFAOYSA-N

Biochem/physiol Actions

CID44216842 is a potent and selective non-competitive allosteric inhibitor of Cdc42 GTPase that does not inhabit Rho and Rac. CID44216842 inhibits Cdc42 dependent filopodia formation and cell migration.
CID44216842 is a potent and selective non-competitive allosteric inhibitor of Cdc42 GTPase.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service