Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5 mg | Check Cart for Availability | € 147,00 |
25 mg | Check Cart for Availability | € 529,00 |
About This Item
Empirical Formula (Hill Notation):
C13H19NO3S
CAS Number:
Molecular Weight:
269.36
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 10 mg/mL, clear
storage temp.
−20°C
SMILES string
[S](=O)(=O)(N2CCCCC2)c1cc(c(cc1)OC)C
InChI
1S/C13H19NO3S/c1-11-10-12(6-7-13(11)17-2)18(15,16)14-8-4-3-5-9-14/h6-7,10H,3-5,8-9H2,1-2H3
InChI key
HDTKLZINZGEPFG-UHFFFAOYSA-N
Application
AA92593 has been used as a selective melanopsin inhibitor.
Biochem/physiol Actions
AA92593 is an opsinamide, a potent antagonist of melanopsin-mediated phototransduction with an IC50 of 665 nM in a Ca2+ flux assay in CHO cells expressing melanopsin (Opn4) and selectivity > 10 μM at 74 common biological targets. AA41612 is believed to act by competing for binding to melanopsin. AA41612 inhibited mouse pupillary light reflex and light aversion in vivo. AA92593 and similar compounds might be useful in alleviating the photophobia associated with migraine.
AA92593 is an opsinamide, a potent antagonist of melanopsin-mediated phototransduction.
AA92593 blocks melanopsin activity and stimulates α-melanocyte-stimulating hormone (α-MSH) expression and induces melanin distribution in the melanophores, which darkens the embryo.
Features and Benefits
This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
1 of 1
This Item | |||
|---|---|---|---|
| description ≥98% (HPLC) | description clone 1F8.1, from mouse | description affinity isolated antibody | description targeting mouse Opn4 |
| assay ≥98% (HPLC) | assay - | assay - | assay - |
| form powder | form - | form buffered aqueous solution | form lyophilized powder |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. −20°C | storage temp. - | storage temp. −20°C | storage temp. −20°C |
| solubility DMSO: 10 mg/mL, clear | solubility - | solubility - | solubility - |
| color white to beige | color - | color - | color - |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




