Skip to Content
Merck
All Photos(1)

Key Documents

SML0838

Sigma-Aldrich

PACMA 31

≥98% (HPLC)

Synonym(s):

N-(2,4-Dimethoxyphenyl)-N-(1-oxo-2-propyn-1-yl)-2-(2-thienyl)glycyl-glycine ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€120.00
25 MG
€336.00

€120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€120.00
25 MG
€336.00

About This Item

Empirical Formula (Hill Notation):
C21H22N2O6S
CAS Number:
Molecular Weight:
430.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€120.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

C#CC(N(C(C1=CC=CS1)C(NCC(OCC)=O)=O)C2=CC=C(OC)C=C2OC)=O

InChI

1S/C21H22N2O6S/c1-5-18(24)23(15-10-9-14(27-3)12-16(15)28-4)20(17-8-7-11-30-17)21(26)22-13-19(25)29-6-2/h1,7-12,20H,6,13H2,2-4H3,(H,22,26)

InChI key

AHOOZADJPNUFOQ-UHFFFAOYSA-N

Biochem/physiol Actions

Both in vivo and in vitro studies reveal that PACMA 31 possesses anti-tumor function, especially observed in ovarian cancer.[1]
PACMA 31 is an orally bioavailable, selective inhibitor (IC50 = 10 μM) of protein disulfide isomerase (PDI), an endoplasmic reticulum protein that regulates oxidative protein folding by catalyzing the breakage and reformation of disulfide bonds. PDI is upregulated in ovarian, and several other cancers. PACMA 31 dose dependently inhibits proliferation of OVCAR-8 in culture and in a tumor xenograft model.
PACMA 31 is an orally bioavailable, selective inhibitor (IC50 = 10 μM) of protein disulfide isomerase (PDI).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Experimental Validation of New PDI Inhibitors with Antiproliferative Activity
Badolato M, et al.
Journal of Chemistry, 2017, 1-10 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service