Skip to Content
Merck
All Photos(1)

Key Documents

SML0812

Sigma-Aldrich

NBD-556

≥98% (HPLC)

Synonym(s):

N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethyl-4-piperidinyl)-ethanediamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€90.20
25 MG
€340.00

€90.20


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€90.20
25 MG
€340.00

About This Item

Empirical Formula (Hill Notation):
C17H24ClN3O2
CAS Number:
Molecular Weight:
337.84
UNSPSC Code:
12352200
NACRES:
NA.77

€90.20


Estimated to ship on30 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)NC(=O)C(=O)NC2CC(NC(C2)(C)C)(C)C

InChI

1S/C17H24ClN3O2/c1-16(2)9-13(10-17(3,4)21-16)20-15(23)14(22)19-12-7-5-11(18)6-8-12/h5-8,13,21H,9-10H2,1-4H3,(H,19,22)(H,20,23)

InChI key

ZKXLQCIOURANAD-UHFFFAOYSA-N

Biochem/physiol Actions

NBD-556 is small molecule mimetic of CD4 that recognizes the HIV-1 envelope protein gp120.
NBD-556 is small molecule mimetic of CD4. NBD-556 recognizes the HIV-1 envelope protein gp120 and induces restructuring of gp120 analogous to CD4 binding. The CD4-induced conformational change in gp120 is necessary for interaction with CCR5.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service