Skip to Content
Merck
All Photos(1)

Key Documents

SML0753

Sigma-Aldrich

PSB-12062

≥98% (HPLC)

Synonym(s):

10-[(4-Methylphenyl)sulfonyl]-10H-phenoxazine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€113.00
25 MG
€355.00

€113.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€113.00
25 MG
€355.00

About This Item

Empirical Formula (Hill Notation):
C19H15NO3S
CAS Number:
Molecular Weight:
337.39
UNSPSC Code:
12352200
NACRES:
NA.77

€113.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N2c3c(cccc3)Oc4c2cccc4)c1ccc(cc1)C

InChI

1S/C19H15NO3S/c1-14-10-12-15(13-11-14)24(21,22)20-16-6-2-4-8-18(16)23-19-9-5-3-7-17(19)20/h2-13H,1H3

InChI key

DHZNMEIBMACSFH-UHFFFAOYSA-N

Application

PSB-12062 has been used as a P2X4 antagonist to pre-treat macrophages in inhibitor screening studies to understand the inflammation process.[1] It has also been used as P2X4 antagonists to pre-treat P2X7-deficient BV-2 cells to elucidate the roles of P2X4.[2]

Biochem/physiol Actions

PSB-12062 is a P2X4 receptor antagonist.
PSB-12062 is a P2X4 receptor antagonist. P2X4 receptors are highly expressed in the central nervous system (CNS), and have been studied as a therapeutic target for neutopathic pain, and treatment of traumatic brain injury, cerebral ischemia, and spinal cord injury. PBS-12062 is a new P2X4-specific antagonist with equal potency against human, rat and mouse receptors (IC50s 1.38, 0.92 and 1.76 μM, respecitively). The compound does not display significant inhibition of human P2X1, P2X2, P2X3 or P2X7 at 10 μM.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Uderhardt et al.
Cell, 177(3), 541-555 (2019-04-09)
Neutrophils are attracted to and generate dense swarms at sites of cell damage in diverse tissues, often extending the local disruption of organ architecture produced by the initial insult. Whether the inflammatory damage resulting from such neutrophil accumulation is an
Kshitija Dhuna et al.
Molecular pharmacology, 95(2), 210-221 (2018-12-14)
We investigated the selectivity of protopanaxadiol ginsenosides from Panax ginseng acting as positive allosteric modulators on P2X receptors. ATP-induced responses were measured in stable cell lines overexpressing human P2X4 using a YOPRO-1 dye uptake assay, intracellular calcium measurements, and whole-cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service