Skip to Content
Merck

Skip To

SML0747

HHQ

≥98% (HPLC)

Synonym(s):

2-Heptyl-4-quinolone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 71,50
50 mg
Check Cart for Availability
€ 292,00
€ 248,20

About This Item

Empirical Formula (Hill Notation):
C16H21NO
CAS Number:
Molecular Weight:
243.34
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
protect from light

€ 71,50


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 5 mg/mL, clear

storage temp.

2-8°C

SMILES string

[nH]1c2c([c](cc1CCCCCCC)=O)cccc2

InChI

1S/C16H21NO/c1-2-3-4-5-6-9-13-12-16(18)14-10-7-8-11-15(14)17-13/h7-8,10-12H,2-6,9H2,1H3,(H,17,18)

InChI key

UYRHHBXYXSYGHA-UHFFFAOYSA-N

General description

HHQ belongs to 2-alkyl-4-quinolone compounds, which are lipophilic and are slightly soluble in water.

Biochem/physiol Actions

HHQ or 2-Heptyl-4-quinolone is a modulator of swarm motility. HHQ can be converted to PQS (Pseudomonas quinolone signal) by the PqsH protein.
HHQ s a modulator of swarm motility.
HHQ stimulates vesicle formation but not as much as PQS.

Other Notes

Light sensitive

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
94398362425
description

≥98% (HPLC)

description

≥96.0% (HPLC)

description

98%

form

powder

form

powder

form

solid

assay

≥98% (HPLC)

assay

≥96.0% (HPLC)

assay

98%

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

solubility

DMSO: 5 mg/mL, clear

solubility

-

solubility

-

storage condition

protect from light

storage condition

-

storage condition

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters