Skip to Content
Merck
All Photos(1)

Key Documents

SML0714

Sigma-Aldrich

FSLLRY-NH2 trifluoroacetate salt

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€113.00

€113.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€113.00

About This Item

Empirical Formula (Hill Notation):
C39H60N10O8 · xC2HF3O2
CAS Number:
Molecular Weight:
796.96 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€113.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

N([C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCN=C(N)N)C(=O)N[C@@H](Cc2ccc(cc2)O)C(=O)N)C(=O)[C@@H](N)Cc1ccccc1

InChI

1S/C39H60N10O8/c1-22(2)17-30(36(55)45-28(11-8-16-44-39(42)43)35(54)46-29(33(41)52)20-25-12-14-26(51)15-13-25)47-37(56)31(18-23(3)4)48-38(57)32(21-50)49-34(53)27(40)19-24-9-6-5-7-10-24/h5-7,9-10,12-15,22-23,27-32,50-51H,8,11,16-21,40H2,1-4H3,(H2,41,52)(H,45,55)(H,46,54)(H,47,56)(H,48,57)(H,49,53)(H4,42,43,44)/t27-,28-,29-,30-,31-,32-/m0/s1

InChI key

KMSCNWHRNILNRJ-JNRWAQIZSA-N

Related Categories

Amino Acid Sequence

Phe-Ser-Leu-Leu-Arg-Tyr-NH2 trifluoroacetate salt

Application

FSLLRY-NH2 trifluoroacetate salt has been used as a protease-activated receptor 2 (PAR2) inhibitor to test its effects on urinary tract infections (UTIs) in a murine model.[1] It has also been used to study the clinical itch datasets from phase 3 clinical trials for moderate-to-severe AD in atopic dermatitis (AD)-associated acute itch flare mice model[2]in rats.[3]

Biochem/physiol Actions

FSLLRY-NH2 is a peptide antagonist of the protease activated receptor 2 (PAR2).
FSLLRY-NH2 is a peptide antagonist of the protease activated receptor 2 (PAR2). The peptide blocks agonist induced itching in rodent models. In cell based assays, the peptide inhibits agonist-induced cardiocyte remodeling, and production of cytokines by endothelial cells.
The ability of FSLLRY-NH2 to decrease visceral hypersensitivity (VH) suggests that PAR2 is involved in the maintenance of VH.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying-Jie Li et al.
Digestive diseases and sciences, 64(5), 1182-1192 (2018-12-19)
Mast cells (MCs), PAR2 and TRPV1, play a key role in the regulation of visceral pain. Several studies have found that probiotics regulate visceral sensitivity. The purpose of the current study was to explore the role of MC-PAR2-TRPV1 in VH
Zonglong Wu et al.
American journal of physiology. Cell physiology, 317(3), C544-C555 (2019-06-27)
The specific regulatory mechanism of bladder urothelial barrier dysfunction after infection with uropathogenic Escherichia coli (UPEC) is still unclear. The cross talk between bladder urothelial cells and mast cells may play an important role during UPEC infection. In this study
Fang Wang et al.
Cell, 184(2), 422-440 (2021-01-16)
Itch is an evolutionarily conserved sensation that facilitates expulsion of pathogens and noxious stimuli from the skin. However, in organ failure, cancer, and chronic inflammatory disorders such as atopic dermatitis (AD), itch becomes chronic, intractable, and debilitating. In addition to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service