Skip to Content
Merck
All Photos(1)

Key Documents

SML0676

Sigma-Aldrich

PY1

≥98% (HPLC)

Synonym(s):

3′-(Diethylamino)-6′-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-spiro[isobenzofuran-1(3H),9′-[9H]xanthen]-3-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€173.00

€173.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€173.00

About This Item

Empirical Formula (Hill Notation):
C30H32BNO5
CAS Number:
Molecular Weight:
497.39
UNSPSC Code:
12352200
NACRES:
NA.77

€173.00


Estimated to ship on27 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, white to dark yellow-brown

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

DPP-Py1 is a ratiometric sensor. It is accurate and reactive with a relatively lower detection limit (1.8 × 10-7 M) than DPP-Py2. It is also stable in various pH buffers than DPP-Py2.[1]
PY1 is a cell-permeable fluorescent H2O2 sensor.
PY1 is a cell-permeable fluorescent sensor that is highly selective for detecting the presence of H2O2 in living cells.

Legal Information

This product may be used for research purposes only. It is not licensed for resale and may only be used by the buyer. This product may not be used and is not licensed for clinical assays, where the results of such assays are provided as a diagnostic service. If a diagnostic or therapeutic use is anticipated, then a license must be requested from the University of California. The availability of such diagnostic and therapeutic use license(s) cannot be guaranteed from the University of California.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diketopyrrolopyrrole-based ratiometric/turn-on fluorescent chemosensors for citrate detection in the near-infrared region by an aggregation-induced emission mechanism.
Hang Y, et al.
Analytical Chemistry, 88(3), 1696-1703 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service