Skip to Content
Merck
All Photos(1)

Key Documents

SML0675

Sigma-Aldrich

α-Methylnorepinephrine

≥95% (HPLC)

Synonym(s):

α-(1-Aminoethyl)-3,4-dihydroxybenzyl alcohol, (-)-α-Methylnorepinephrine, 4-(2-Amino-1-hydroxypropyl)-1,2-benzenediol, L-4-(2-Amino-1-hydroxypropyl)-1,2-benzenediol, (-)-Nordefrin, (-)3,4-Dihydroxynorephedrine, Corbadrine, Levonordefrin

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€71.60
50 MG
€279.00

€71.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€71.60
50 MG
€279.00

About This Item

Empirical Formula (Hill Notation):
C9H13NO3
CAS Number:
Molecular Weight:
183.20
UNSPSC Code:
12352200
NACRES:
NA.77

€71.60


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

N[C@H]([C@H](O)c1cc(c(cc1)O)O)C

InChI

1S/C9H13NO3/c1-5(10)9(13)6-2-3-7(11)8(12)4-6/h2-5,9,11-13H,10H2,1H3/t5-,9-/m0/s1

InChI key

GEFQWZLICWMTKF-CDUCUWFYSA-N

Biochem/physiol Actions

α-Methylnorepinephrine (αMNE) is an α2-adrenergic agonist and is useful in cardiopulmonary resuscitation (CPR) during myocardial dysfunction especially in animals.[1] αMNE improves water absorption in the jejunum and is blood-brain barrier impermeable.[2]
α-Methylnorepinephrine is a sympathomimetic amine and vasocinstrictor.
α-Methylnorepinephrine is a sympathomimetic amine and vasoconstrictor. α-Methylnorepinephrine is used clinically in dentistry in local anesthetic solutions.

Features and Benefits

This compound is featured on the α2-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of alpha-methylnorepinephrine, an alpha 2-adrenergic agonist, on jejunal absorption in neurally intact conscious dog
Herkes SM, et al.
Digestive Diseases and Sciences, 38(9), 1645-1650 (1993)
alpha-Methylnorepinephrine, a selective alpha2-adrenergic agonist for cardiac resuscitation
Sun S, et al.
Journal of the American College of Cardiology, 37(3), 951-956 (2001)

Articles

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service