Skip to Content
Merck
All Photos(1)

Key Documents

SML0668

Sigma-Aldrich

Calhex 231 hydrochloride

≥98% (HPLC)

Synonym(s):

4-Chloro-N-[(1S,2S)-2-[[(1R)-1-(1-naphthalenyl)ethyl]amino]cyclohexyl]-benzamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€144.00
25 MG
€419.00

€144.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€144.00
25 MG
€419.00

About This Item

Empirical Formula (Hill Notation):
C25H27ClN2O · HCl
CAS Number:
Molecular Weight:
443.41
UNSPSC Code:
12352200
NACRES:
NA.77

€144.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

Application

Calhex 231 hydrochloride has been used:
  • as a calcium-sensing receptor (CaSR) antagonist: to test its impact on hearing in rats[1]
  • to test its effect on blood pressure levels spontaneous hypertensive rats (SHRs)[2]
  • to test its effect on acoustic response bias in zebrafish[3]
  • in Caco-2 cells to test its effect on, overexpression of fibroblast growth factor-23 (FGF-23 )[4]

Biochem/physiol Actions

Calhex 231 is a negative allosteric modulator of the calcium sensing receptor CaSR.
Calhex 231 is a negative allosteric modulator of the calcium sensing receptor CaSR. Calhex 231 blocks calcium-induced accumulation of inositol phosphate with an IC50 of 0.36 μM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiya Minakata et al.
Frontiers in molecular neuroscience, 12, 175-175 (2019-08-06)
Maintaining Ca2+ homeostasis in lymphatic fluids is necessary for proper hearing. Despite its significance, the mechanisms that maintain the cochlear lymphatic Ca2+ concentrations within a certain range are not fully clarified. We investigated the functional expression of calcium-sensing receptor (CaSR)
Xin Zhang et al.
Mediators of inflammation, 2019, 6847087-6847087 (2019-03-25)
Increasing evidence suggests that the NLRP3 (nucleotide oligomerization domain-like receptor family, pyrin domain containing 3) inflammasome participates in cardiovascular diseases. However, its role and activation mechanism during hypertension remains unclear. In this study, we tested the role and mechanism of
Kannikar Wongdee et al.
Archives of biochemistry and biophysics, 657, 15-22 (2018-09-16)
Long-term high-calcium intake and intestinal calcium hyperabsorption are hazardous to the body. It is hypothesized that enterocytes possess mechanisms for preventing superfluous calcium absorption, including secretion of negative regulators of calcium absorption and utilization of calcium-sensing receptor (CaSR) to detect
Roshan A Jain et al.
Current biology : CB, 28(9), 1357-1369 (2018-04-24)
Animals continuously integrate sensory information and select contextually appropriate responses. Here, we show that zebrafish larvae select a behavioral response to acoustic stimuli from a pre-existing choice repertoire in a context-dependent manner. We demonstrate that this sensorimotor choice is modulated
Pamela Mattar et al.
International journal of molecular sciences, 21(20) (2020-10-21)
Autophagy is upregulated in adipose tissue (AT) from people with obesity. We showed that activation of the calcium-sensing receptor (CaSR) elevates proinflammatory cytokines through autophagy in preadipocytes. Our aim is to understand the role of CaSR on autophagy in AT

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service