Skip to Content
Merck
All Photos(1)

Key Documents

SML0660

Sigma-Aldrich

GSK2578215A

≥98% (HPLC)

Synonym(s):

5-(2-Fluoro-4-pyridinyl)-2-(phenylmethoxy)-N-3-pyridinyl-benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€113.00
25 MG
€340.00

€113.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€113.00
25 MG
€340.00

About This Item

Empirical Formula (Hill Notation):
C24H18FN3O2
CAS Number:
Molecular Weight:
399.42
UNSPSC Code:
12352200
NACRES:
NA.77

€113.00


Estimated to ship on23 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1nccc(c1)c2cc(c(cc2)OCc4ccccc4)C(=O)Nc3cnccc3

InChI

1S/C24H18FN3O2/c25-23-14-19(10-12-27-23)18-8-9-22(30-16-17-5-2-1-3-6-17)21(13-18)24(29)28-20-7-4-11-26-15-20/h1-15H,16H2,(H,28,29)

InChI key

WCIGMFCFPXZRMQ-UHFFFAOYSA-N

Biochem/physiol Actions

GSK2578215A is a potent inhibitor of leucine-rich repeat kinase-2 (LRRK2) also known as dardarin or PARK8, a kinase with mutations linked to Parkinson′s disease. Because the most common mutation, G2019S, enhances LRRK2 kinase activity, it is hoped that LRRK2 inhibitors may be useful in treating the disease. GSK2578215A inhibited both wild-type and G2019S mutant LRRK2 kinase activity with IC50s of 10.9 and 8.9 nM respectively. It has good blood-brain barrier (BBB) permeability with a high ratio of brain to plasma distribution in mice.
GSK2578215A is a potent inhibitor of leucine-rich repeat kinase-2 (LRRK2).
GSK2578215A stimulates mitochondrial fragmentation, autophagy and mitophagy.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The LRRK2 inhibitor GSK2578215A induces protective autophagy in SH-SY5Y cells: involvement of Drp-1-mediated mitochondrial fission and mitochondrial-derived ROS signaling
Saez-Atienzar S, et al.
Cell Death & Disease, 5(8), e1368-e1368 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service