Skip to Content
Merck
All Photos(1)

Key Documents

SML0604

Sigma-Aldrich

SB-399885 hydrochloride

≥98% (HPLC)

Synonym(s):

N-(3,5-Dichloro-2-methoxyphenyl)-4-methoxy-3-(1-piperazinyl)-benzenesulfonamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€142.00

€142.00


Estimated to ship on03 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€142.00

About This Item

Empirical Formula (Hill Notation):
C18H21Cl2N3O4S · HCl
CAS Number:
Molecular Weight:
482.81
UNSPSC Code:
12352200
NACRES:
NA.77

€142.00


Estimated to ship on03 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 15 mg/mL, clear

storage temp.

room temp

SMILES string

[S](=O)(=O)(Nc3c(c(cc(c3)Cl)Cl)OC)c1cc(c(cc1)OC)N2CCNCC2

InChI

1S/C18H21Cl2N3O4S/c1-26-17-4-3-13(11-16(17)23-7-5-21-6-8-23)28(24,25)22-15-10-12(19)9-14(20)18(15)27-2/h3-4,9-11,21-22H,5-8H2,1-2H3

InChI key

ATKZKAYWARYLBW-UHFFFAOYSA-N

Biochem/physiol Actions

SB-399885 is a potent, selective, and orally active 5-HT6 receptor antagonist with greater than 200 fold selectivity over other 5-HT receptors. SB-399885 is brain penetrant and displays antidepressant and anxiolytic activity in rodent models.
SB-399885 is a potent, selective, and orally active 5-HT6 receptor antagonist; brain penetrant.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ao Li et al.
CNS neuroscience & therapeutics, 27(8), 941-950 (2021-05-12)
General anesthesia has been widely applied in surgical or nonsurgical medical procedures, but the mechanism behind remains elusive. Because of shared neural circuits of sleep and anesthesia, whether serotonergic system, which is highly implicated in modulation of sleep and wakefulness

Articles

We offer many products related to serotonin receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service