Skip to Content
Merck
All Photos(1)

Documents

SML0555

Sigma-Aldrich

ML289

≥98% (HPLC)

Synonym(s):

VU0463597, [(3R)-3-(Hydroxymethyl)-1-piperidinyl][4-[2-(4-methoxyphenyl)ethynyl]phenyl]-methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23NO3
CAS Number:
Molecular Weight:
349.42
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C22H23NO3/c1-26-21-12-8-18(9-13-21)5-4-17-6-10-20(11-7-17)22(25)23-14-2-3-19(15-23)16-24/h6-13,19,24H,2-3,14-16H2,1H3/t19-/m1/s1

InChI key

VSLWUPHHCFQTDB-LJQANCHMSA-N

Biochem/physiol Actions

ML289 (VU0463597) is a potent, selective and centrally penetrant negative allosteric modifier (NAM) of metabotropic glutamate receptor 3 (mGlu3). ML289 is >15-fold selective vs mGlu2 with an IC50 value of 0.66 μM.

Features and Benefits

This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service