Skip to Content
Merck
All Photos(1)

Key Documents

SML0542

Sigma-Aldrich

SR 27417

≥98% (HPLC)

Synonym(s):

Faropafant, N,N-Dimethyl-N′-(3-pyridinylmethyl)-N′-[4-[2,4,6-tris(1-methylethyl)phenyl]-2-thiazolyl]-1,2-ethanediamine, N1,N1-Dimethyl-N2-(3-pyridinylmethyl)-N2-[4-[2,4,6-tris(1-methylethyl)phenyl]-2-thiazolyl]-1,2-ethanediamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€131.00
25 MG
€373.00

€131.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€131.00
25 MG
€373.00

About This Item

Empirical Formula (Hill Notation):
C28H40N4S
CAS Number:
Molecular Weight:
464.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€131.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution, warmed)

storage temp.

2-8°C

SMILES string

CC(C)c1cc(C(C)C)c(-c2csc(n2)N(CCN(C)C)Cc3cccnc3)c(c1)C(C)C

InChI

1S/C28H40N4S/c1-19(2)23-14-24(20(3)4)27(25(15-23)21(5)6)26-18-33-28(30-26)32(13-12-31(7)8)17-22-10-9-11-29-16-22/h9-11,14-16,18-21H,12-13,17H2,1-8H3

InChI key

VVBFISAUNSXQGZ-UHFFFAOYSA-N

Biochem/physiol Actions

SR 27417 is a long-acting, highly potent, specific competitive platelet-activating factor (PAF) receptor antagonist.
SR 27417 is a selective, highly potent, competitive platelet-activating factor (PAF) receptor antagonist.

Features and Benefits

This compound is featured on the PAF Receptor page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N M Thielman et al.
The Journal of clinical investigation, 99(8), 1999-2004 (1997-04-15)
Cholera toxin (CT)-induced intestinal secretion and Chinese hamster ovary cell (CHO) elongation involves cyclic adenosine monophosphate and protein synthesis-dependent prostaglandin formation. We previously reported inhibition of CT-induced intestinal secretion and CHO elongation by platelet-activating factor (PAF) receptor antagonists and secretion
J M Herbert et al.
Journal of lipid mediators and cell signalling, 15(2), 115-123 (1997-01-01)
SR 27417, a potent PAF receptor antagonist, inhibited in a dose-dependent manner the hypotensive effect of PAF in rats. It protected rats with an ED50 value of 6 +/- 2 micrograms/kg (n = 6), when given i.v., 1 min before
D J Evans et al.
American journal of respiratory and critical care medicine, 156(1), 11-16 (1997-07-01)
Platelet-activating factor (PAF) is a lipid-derived mediator that has been implicated in the pathophysiology of airway inflammation in asthma. Its actions include chemotaxis and activation of inflammatory cells, particularly eosinophils. Inhaled PAF causes bronchoconstriction and increased airway responsiveness in human
S O Heard et al.
Journal of critical care, 10(1), 7-14 (1995-03-01)
To determine if platelet-activating factor (PAF) is a key mediator of lipopolysaccharide (LPS)-induced myocardial depression in guinea pigs. Hartley guinea pigs of either sex received intraperitoneal (IP) injections of either vehicle (n = 45) or one of three chemically dissimilar
H C Castro-Faria-Neto et al.
Planta medica, 61(2), 106-112 (1995-04-01)
The pharmacological profile of a novel specific platelet-activating factor (PAF) receptor antagonist-yangambin-isolated from the Brazilian plant Ocotea duckei Vattimo (Lauraceae), was investigated in the pentobarbitone-anaesthetized rabbit. The i.v. administration of PAF (0.03-3.0 microgram kg-1) induced marked but reversible hypotensive effects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service