Skip to Content
Merck
All Photos(1)

Key Documents

SML0518

Sigma-Aldrich

NSC405020

≥98% (HPLC)

Synonym(s):

3,4-Dichloro-N-(1-methylbutyl)-benzamide, 3,4-Dichloro-N-(pentan-2-yl)benzamide, NSC 405020

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€117.00
25 MG
€366.00

€117.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€117.00
25 MG
€366.00

About This Item

Empirical Formula (Hill Notation):
C12H15Cl2NO
CAS Number:
Molecular Weight:
260.16
UNSPSC Code:
12352200
NACRES:
NA.77

€117.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

room temp

SMILES string

Clc1c(ccc(c1)C(=O)NC(CCC)C)Cl

InChI

1S/C12H15Cl2NO/c1-3-4-8(2)15-12(16)9-5-6-10(13)11(14)7-9/h5-8H,3-4H2,1-2H3,(H,15,16)

InChI key

ARDYECYBETXQFD-UHFFFAOYSA-N

Biochem/physiol Actions

NSC405020 is a PEX inhibitor targeting the PEX domain of MT1-MMP that repressed tumor growth and caused a fibrotic DPEX-like tumor phenotype in vivo. NSC405020 does not inhibit the catalytic activity of MMP-2.
NSC405020 is a PEX inhibitor targeting the PEX domain of MT1-MMP.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Rolland et al.
Journal of molecular cell biology (2022-10-23)
Understanding how certain animals are capable of regenerating their hearts will provide much needed insights into how this process can be induced in humans in order to reverse the damage caused by myocardial infarction. Currently, it is becoming increasingly evident

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service