Skip to Content
Merck
All Photos(1)

Key Documents

SML0495

Sigma-Aldrich

Ro 25-6981 maleate salt

≥98% (HPLC)

Synonym(s):

1-Piperidinepropanol, α-(4-hydroxyphenyl)-β-methyl-4-(phenylmethyl)-, ( αR,βS)-, (2Z)-2-butenedioate (1:1), [R-(R*,S*)]-α-(4-Hydroxyphenyl)-β-methyl-4-(phenylmethyl)-1-piperidinepropanol maleate salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€128.00
25 MG
€327.00

€128.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€128.00
25 MG
€327.00

About This Item

Empirical Formula (Hill Notation):
C22H29NO2 · C4H4O4
CAS Number:
Molecular Weight:
455.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€128.00


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +15 to +22°, c = 1 in methanol

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL (clear solution, warmed)

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O.C[C@@H](CN1CCC(CC1)Cc2ccccc2)[C@@H](O)c3ccc(O)cc3

InChI

1S/C22H29NO2.C4H4O4/c1-17(22(25)20-7-9-21(24)10-8-20)16-23-13-11-19(12-14-23)15-18-5-3-2-4-6-18;5-3(6)1-2-4(7)8/h2-10,17,19,22,24-25H,11-16H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t17-,22+;/m0./s1

InChI key

FYJZEHCQSUBZDY-SEELMCCHSA-N

Application

Ro 25-6981 maleate salt has been used as a selective N-methyl D-aspartate receptor subtype 2B (NR2B) blocker to study its effects on patch-clamp recordings of N-methyl D-aspartate receptor subtype 2A (NR2A)-containing N-methyl D-aspartate receptor (NMDAR) excitatory postsynaptic currents (EPSCs).[1]

Biochem/physiol Actions

Ro 25-6981 maleate salt is a potent and selective antagonist of NMDA glutamate receptors containing the NR2B subunit with IC50 values of .009 μM for NR2B vs. 52 μM for NR2A subunits. Ro 25-6981 has been shown to have neuroprotectant effects and to reduce inflammatory and neuropathic pain in rodent models.
Ro 25-6981 maleate salt is a potent and selective antagonist of NMDA glutamate receptors containing the NR2B subunit.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydroxysafflor yellow A protects neurons from excitotoxic death through inhibition of NMDARs
Wang X, et al.
ASN Neuro, 8(2), 1759091416642345-1759091416642345 (2016)

Articles

We offers many products related to ion channel family glutamate receptors for your research needs.

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service