Skip to Content
Merck
All Photos(1)

Key Documents

SML0483

Sigma-Aldrich

DpC

≥98% (HPLC)

Synonym(s):

Di-2-pyridylketone-4-cyclohexyl-4-methyl-3-thiosemicarbazone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€156.00
25 MG
€586.00

€156.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€156.00
25 MG
€586.00

About This Item

Empirical Formula (Hill Notation):
C19H23N5S
CAS Number:
Molecular Weight:
353.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€156.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution)

storage temp.

−20°C

SMILES string

CN(C1CCCCC1)C(=S)N\N=C(\c2ccccn2)c3ccccn3

InChI

1S/C19H23N5S/c1-24(15-9-3-2-4-10-15)19(25)23-22-18(16-11-5-7-13-20-16)17-12-6-8-14-21-17/h5-8,11-15H,2-4,9-10H2,1H3,(H,23,25)

InChI key

GNLZNQJBZNOUBM-UHFFFAOYSA-N

Biochem/physiol Actions

DpC is an iron-chelating agent.
DpC is an iron-chelating agent. In four separate human pancreatic tumor cell lines, DpC significantly increased the expression and phosphorylation of the growth and metastasis suppressor, N-myc down-stream regulated gene 1 (NDRG1) and the cdk inhibitpr p21CIP1/WAF. DpC completely blocked PANC-1 tumor growth in mouse xenografts.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ye-Xing Xu et al.
Oncology letters, 15(5), 7999-8004 (2018-05-10)
The present study aimed to investigate the antitumor efficacy of di-2-pyridylketone-4-cyclohexyl-4-methyl-3-thiosemicarbazone (DpC) and di-2-pyridylketone-4,4,-dimethyl-3-thiosemicarbazone (Dp44mT) on head and neck squamous cell carcinoma (HNSCC) cells. The proliferation and apoptosis of HNSCC cells treated with the iron chelators DpC and Dp44mT were
Sang Y Lee et al.
Anticancer research, 41(6), 2795-2804 (2021-06-05)
Neuroblastoma is the most common childhood extracranial solid malignancy. Although cancer cells need iron and lipids for active cell division, possible links between iron and lipid metabolism in neuroblastomas have not been studied. We evaluated the levels and association between
Zaklina Kovacevic et al.
The Journal of biological chemistry, 291(3), 1029-1052 (2015-11-05)
N-MYC downstream-regulated gene-1 (NDRG1) is a potent growth and metastasis suppressor that acts through its inhibitory effects on a wide variety of cellular signaling pathways, including the TGF-β pathway, protein kinase B (AKT)/PI3K pathway, RAS, etc. To investigate the hypothesis
Ruxing Xi et al.
Molecular pharmacology, 91(5), 499-517 (2017-03-10)
Tumor necrosis factor α (TNFα) plays a vital role in cancer progression as it is associated with inflammation and promotion of cancer angiogenesis and metastasis. The effects of TNFα are mediated by its downstream target, the oncogene lysine-rich CEACAM1 coisolated
Wensheng Liu et al.
Oncotarget, 6(11), 8851-8874 (2015-04-11)
N-myc downstream regulated gene-1 (NDRG1) is a potent metastasis suppressor that plays a key role in regulating signaling pathways involved in mediating cancer cell invasion and migration, including those derived from prostate, colon, etc. However, the mechanisms and molecular targets

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service