Skip to Content
Merck
All Photos(1)

Key Documents

SML0442

Sigma-Aldrich

ML239

≥98% (HPLC)

Synonym(s):

(2,4,6-Trichloro-phenoxy)-acetic acid (1H-pyrrol-2-ylmethylene)-hydrazide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€97.60

€97.60


Estimated to ship on04 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€97.60

About This Item

Empirical Formula (Hill Notation):
C13H10Cl3N3O2
CAS Number:
Molecular Weight:
346.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€97.60


Estimated to ship on04 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

Clc1cc(Cl)c(OCC(=O)N\N=C\c2ccc[nH]2)c(Cl)c1

InChI

1S/C13H10Cl3N3O2/c14-8-4-10(15)13(11(16)5-8)21-7-12(20)19-18-6-9-2-1-3-17-9/h1-6,17H,7H2,(H,19,20)/b18-6+

InChI key

NVEDPFICKAIHKD-NGYBGAFCSA-N

Application

ML239 has been used as a proble in cellular lipid profiling.[1]

Biochem/physiol Actions

ML239 decreases the saturated and monounsaturated lysophosphatidylethanolamines in lung cancer cells and is toxic to breast cancer cells.[1] ML239 may elicit toxicity by modulating the nuclear factor−κB (NF-κB) associated pathways.[2]
ML239 is an inhibitor of breast cancer stem cells found from a screen using stem cell-like human mammary epithelial cells (HMLE). ML239 inhibited cancer stem cells with an IC50 of 1.16 μM with 24-fold selectivity against the control cell line.
ML239 is an inhibitor of breast cancer stem cells.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phenotypic High-Throughput Screening Elucidates Target Pathway in Breast Cancer Stem Cell-Like Cells
Carmody LC, et al.
Journal of Biomolecular Screening, 17(9), 1204-1210 (2012)
Correlating chemical sensitivity and basal gene expression reveals mechanism of action
Rees MG, et al.
Nature chemical biology, 12(2), 109-109 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service