Skip to Content
Merck
All Photos(1)

Key Documents

SML0391

Sigma-Aldrich

SB-268262

≥98% (HPLC)

Synonym(s):

(+/-)-SB-273779, N-Methyl-N-(2-methylphenyl)-3-nitro-4-(2-thiazolylsulfinyl)-benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€76.30
25 MG
€299.00

€76.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€76.30
25 MG
€299.00

About This Item

Empirical Formula (Hill Notation):
C18H15N3O4S2
CAS Number:
Molecular Weight:
401.46
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

€76.30


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

CN(C(=O)c1ccc(c(c1)[N+]([O-])=O)S(=O)c2nccs2)c3ccccc3C

InChI

1S/C18H15N3O4S2/c1-12-5-3-4-6-14(12)20(2)17(22)13-7-8-16(15(11-13)21(23)24)27(25)18-19-9-10-26-18/h3-11H,1-2H3

InChI key

YXHLPJQQPOMPGE-UHFFFAOYSA-N

Biochem/physiol Actions

SB-268262 is a selective non-peptide antagonist at the CGRP1 receptor.
SB-268262 is a selective non-peptide antagonist at the CGRP1 receptor. SB-268262 inhibits CGRP binding, and CGRP-mediated stimulation of adenylyl cyclase activity in human neuroblastoma cells with IC50 values of 240 and 830 nM, respectively.

Features and Benefits

This compound is featured on the Calcitonin Gene-Related Peptides page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service