Skip to Content
Merck
All Photos(1)

Key Documents

SML0369

Sigma-Aldrich

CID-2011756

≥98% (HPLC)

Synonym(s):

5-(3-Chlorophenyl)-N-[4-(morpholin-4-ylmethyl)phenyl]furan-2-carboxamide, CID 2011756

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€105.00
25 MG
€270.00

€105.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€105.00
25 MG
€270.00

About This Item

Empirical Formula (Hill Notation):
C22H21ClN2O3
CAS Number:
Molecular Weight:
396.87
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€105.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL (clear solution)

storage temp.

room temp

SMILES string

Clc1cccc(c1)-c2ccc(o2)C(=O)Nc3ccc(CN4CCOCC4)cc3

InChI

1S/C22H21ClN2O3/c23-18-3-1-2-17(14-18)20-8-9-21(28-20)22(26)24-19-6-4-16(5-7-19)15-25-10-12-27-13-11-25/h1-9,14H,10-13,15H2,(H,24,26)

InChI key

XQJWTJLJEYIUDZ-UHFFFAOYSA-N

Biochem/physiol Actions

CID-2011756 is a cell-active ATP competitive and specific PKD1 inhibitor that inhibits phorbol ester-induced endogenous PKD1 activation in LNCaP prostate cancer cells.
CID-2011756 is a cell-active ATP competitive and specific PKD1 inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service