Skip to Content
Merck
All Photos(1)

Key Documents

SML0298

Sigma-Aldrich

PSNCBAM-1

≥98% (HPLC)

Synonym(s):

N-(4-Chlorophenyl)-N′-[3-[6-(1-pyrrolidinyl)-2-pyridinyl]phenyl]-urea

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€141.00
25 MG
€356.00

€141.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€141.00
25 MG
€356.00

About This Item

Empirical Formula (Hill Notation):
C22H21ClN4O
CAS Number:
Molecular Weight:
392.88
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€141.00


Estimated to ship on18 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

Clc1ccc(NC(=O)Nc2cccc(c2)-c3cccc(n3)N4CCCC4)cc1

InChI

1S/C22H21ClN4O/c23-17-9-11-18(12-10-17)24-22(28)25-19-6-3-5-16(15-19)20-7-4-8-21(26-20)27-13-1-2-14-27/h3-12,15H,1-2,13-14H2,(H2,24,25,28)

InChI key

HDAYFSFWIPRJSO-UHFFFAOYSA-N

Biochem/physiol Actions

PSNCBAM-1 is an allosteric, non-competitive antagonist of CB-1.
PSNCBAM-1 is an allosteric, non-competitive antagonist of CB-1. The compound blocks the effects of WIN 55212-2 and CP55940, and reduces food intake and weight gain in rats.

Features and Benefits

This compound is featured on the Cannabinoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas F Gamage et al.
Neuropharmacology, 125, 365-375 (2017-08-15)
While allosteric modulators of the cannabinoid type-1 receptor (CB

Articles

We offer many products related to cannabinoid receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service