Skip to Content
Merck

Skip To

SML0263

PF-04217903

≥98% (HPLC)

Synonym(s):

4-[1-(6-Quinolinylmethyl)-1H-1,2,3-triazolo[4,5-b]pyrazin-6-yl]-1H-pyrazole-1-ethanol methanesulfonate (1:1)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 355,00

About This Item

Empirical Formula (Hill Notation):
C19H16N8O·CH3SO3H
CAS Number:
Molecular Weight:
468.49
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder

€ 355,00


Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥5 mg/mL (warmed)

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.OCCn1cc(cn1)-c2cnc3nnn(Cc4ccc5ncccc5c4)c3n2

InChI

1S/C19H16N8O.CH4O3S/c28-7-6-26-12-15(9-22-26)17-10-21-18-19(23-17)27(25-24-18)11-13-3-4-16-14(8-13)2-1-5-20-16;1-5(2,3)4/h1-5,8-10,12,28H,6-7,11H2;1H3,(H,2,3,4)

InChI key

HBEMHKVWZJTVOC-UHFFFAOYSA-N

Gene Information

human ... MET(4233)

Application

PF-04217903 has been used as tyrosine-protein kinase Met (C-Met) selective inhibitor in Madin-Darby Canine kidney (MDCK) cells and NT2D1 non-seminoma cells.

Biochem/physiol Actions

PF-04217903 is a c-Met inhibitor.
PF-04217903 is a highly selective, potent inhibitor of the hepatocyte growth factor receptor c-Met. PF-04217903 inhibits endogenous, wild type c-Met in A549 human lung carcinoma cells with an IC50 of 4.8 nM. The compund displays 1000-fold selectivity against a panel of 208 other kinases.
PF-04217903 is an ATP-competitive inhibitor. It elicits antiangiogenic functionality. PF-04217903 inhibits c-Met phosphorylation in xenograft models leading to partial tumor growth suppression.

Features and Benefits

This compound is featured on the Met page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y4895SML2824SML1276
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: ≥5 mg/mL (warmed)

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

white to beige


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters