Skip to Content
Merck
All Photos(1)

Key Documents

SML0228

Sigma-Aldrich

Rc-Ome

≥98% (HPLC)

Synonym(s):

(2E)-3-(2-hydroxy-5-methoxyphenyl)-1-(4-methoxyphenyl)-2-Propen-1-one, trans-6,4′-dimethoxyretrochalcone

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€182.00

€182.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€182.00

About This Item

Empirical Formula (Hill Notation):
C17H16O4
CAS Number:
Molecular Weight:
284.31
UNSPSC Code:
12352202
NACRES:
NA.77

€182.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, light to dark yellow

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

Application

Rc-Ome may be used in GABAA-mediated cell signaling studies.

Biochem/physiol Actions

Rc-OMe is a GABAA PAM
Rc-OMe is a GABAA PAM that distinguishes α1β2γ2s from α1β2 receptors. The compound has very weak potentiating effects on α1β2, but potentiates GABA-elicited currents of α(123)β2γ2s receptors, all with low μM EC50s. Rc-OMe acts at a binding site that is distinct from BDZ, pentobarbital and loreclezole modulatory sites.

Features and Benefits

This compound is featured on the GABAA Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to GABAA receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service