Skip to Content
Merck
All Photos(1)

Key Documents

SML0214

Sigma-Aldrich

SAMT-247

≥98% (HPLC)

Synonym(s):

3-(S-Acetylthiosalicyloyl)aminopropamide, N-[2-Acetylthiobenzoyl]-β-alaninamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€82.90

€82.90


Check Cart for Availability


Select a Size

Change View
5 MG
€82.90

About This Item

Empirical Formula (Hill Notation):
C12H14N2O3S
CAS Number:
Molecular Weight:
266.32
UNSPSC Code:
12352200
NACRES:
NA.77

€82.90


Check Cart for Availability

Assay

≥98% (HPLC)

form

powder

color

white to light yellow

solubility

H2O: ≥2 mg/mL at 60 °C

storage temp.

2-8°C

Application

SAMT-247 may be used in HIV-1 related studies.

Biochem/physiol Actions

HIV inhibitor by modifying the NCp7 region of Gag in infected cells
SAMT-247 inhibits HIV by modifying the nucleocapsid NCp7 region of Gag in infected cells, blocking Gag processing and reducing infectivity.
SAMT-247 is a microbicide that decreases the viral infectivity and infectivity of the progeny virus in HIV-1 infections.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cecilia Cheng-Mayer et al.
AIDS (London, England), 25(15), 1833-1841 (2011-07-14)
Development of an effective vaccine or topical compound to prevent HIV transmission remains a major goal for control of the AIDS pandemic. Using a nonhuman primate model of heterosexual HIV-1 transmission, we tested whether a topical microbicide that reduces viral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service