Skip to Content
Merck
All Photos(1)

Key Documents

SML0125

Sigma-Aldrich

CPA 1 hydrate

≥98% (HPLC)

Synonym(s):

NSC 623721 hydrate, Naphtho[2,1-b:7,6-b′]difuran-2,8-dicarboxylic acid hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€150.00

€150.00


Estimated to ship on07 May 2025



Select a Size

Change View
5 MG
€150.00

About This Item

Empirical Formula (Hill Notation):
C16H8O6 · xH2O
CAS Number:
Molecular Weight:
296.23 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€150.00


Estimated to ship on07 May 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

gray-brown

solubility

DMSO: ≥15 mg/mL

storage temp.

2-8°C

SMILES string

O.OC(=O)c1cc2cc3ccc4oc(cc4c3cc2o1)C(O)=O

InChI

1S/C16H8O6.H2O/c17-15(18)13-4-8-3-7-1-2-11-10(6-14(21-11)16(19)20)9(7)5-12(8)22-13;/h1-6H,(H,17,18)(H,19,20);1H2

InChI key

HKVITMNNEVXIGB-UHFFFAOYSA-N

Biochem/physiol Actions

CPA 1 is a potent, selective, ATP-competitive dual inhibitor of protein kinase casein kinase 2 (CK2) and proviral insertion Moloney virus kinases (Pim kinases). CPA 1 is selective for CK2 and Pim kinases (Pim-1, Pim-2, and Pim-3) when evaluated against a panel of 112 protein kinases.
Potent, selective, ATP-competitive dual inhibitor of protein kinase casein kinase 2 (CK2) and proviral insertion Moloney virus kinases (Pim kinases)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service