Skip to Content
Merck
All Photos(1)

Key Documents

SML0047

Sigma-Aldrich

MoTP

≥98% (HPLC)

Synonym(s):

33N14, 4-(4-Morpholinobutylthio)phenol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€62.70
25 MG
€355.00

€62.70


Estimated to ship on14 May 2025



Select a Size

Change View
5 MG
€62.70
25 MG
€355.00

About This Item

Empirical Formula (Hill Notation):
C14H21NO2S
CAS Number:
Molecular Weight:
267.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€62.70


Estimated to ship on14 May 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥80 mg/mL

storage temp.

−20°C

SMILES string

Oc1ccc(SCCCCN2CCOCC2)cc1

InChI

1S/C14H21NO2S/c16-13-3-5-14(6-4-13)18-12-2-1-7-15-8-10-17-11-9-15/h3-6,16H,1-2,7-12H2

InChI key

YHKNLUTUXZDOQI-UHFFFAOYSA-N

Application

MoTP may be used in cell signaling and differentiation studies.

Biochem/physiol Actions

Melanocytotoxic; reversibly blocks the specification of neural crest cells into melanocytes
MoTP (also known as 33N14) reversibly blocks the specification of neural crest cells into melanocytes in zebrafish studies. Differentiation of neural crest cell-derived melanoblasts proceeds to late developmental stages, but they fail to reach the terminal stage of melanin production and die. Melanocytotoxicity is dependent on tyrosinase activity which convert it into a toxic form. MoTP specifically ablates only cells that express tyrosinase at high levels, which in the embryo are limited to melanoblasts and newly formed melanocytes. MoTP should be a useful tool for elucidating melanocyte stem cell regeneration, recruitment, and maintenance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kleio Petratou et al.
PloS one, 16(1), e0244794-e0244794 (2021-01-14)
Understanding how fate specification of distinct cell-types from multipotent progenitors occurs is a fundamental question in embryology. Neural crest stem cells (NCSCs) generate extraordinarily diverse derivatives, including multiple neural, skeletogenic and pigment cell fates. Key transcription factors and extracellular signals
Li Liu et al.
Biomolecules, 10(9) (2020-09-24)
It has been reported that 5-hydroxytryptamine (5-HT) is related to melanogenesis in mice and melanoma cells. However, the underlying mechanisms of 5-HT in regulating pigmentation remains unknown. In this study, we aim to clarify the regulatory mechanism of 5-HT in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service