Skip to Content
Merck
All Photos(1)

Key Documents

SMB01068

Sigma-Aldrich

Licoricidin

≥90% (LC/MS-ELSD)

Synonym(s):

(+)-Licoricidin, 7-O-Demethyllicorisoflavan A, Licorisoflavan B

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€306.00

€306.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€306.00

About This Item

Empirical Formula (Hill Notation):
C26H32O5
CAS Number:
Molecular Weight:
424.53
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

€306.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

424.53

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1C[C@H](Cc3c1cc(c(c3OC)CC=C(C)C)O)c2c(c(c(cc2)O)CC=C(C)C)O

InChI

1S/C26H32O5/c1-15(2)6-8-19-22(27)11-10-18(25(19)29)17-12-21-24(31-14-17)13-23(28)20(26(21)30-5)9-7-16(3)4/h6-7,10-11,13,17,27-29H,8-9,12,14H2,1-5H3/t17-/m0/s1

InChI key

GBRZTUJCDFSIHM-KRWDZBQOSA-N

General description

Licoricidin, also known as Licorisoflavan B, a hydroxyisoflavan and an aromatic ether, is a bioactive natural compound commonly derived from Glycyrrhiza Sp. (G. uralensis, G. inflata, and G. aspera) plants. Current research indicates that this metabolite is an inhibitor and may possess diverse biological activities, including anti-inflammatory, antitumor, antibacterial, antiangiogenic and antimetastatic properties.

Application

Licoricidin is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Licoricidin is a potent anti-metastatic agent, effectively reducing the metastatic and invasive capabilities of malignant prostate cancer cells. Additionally, both Licoricidin and Licorisoflavan A exhibit strong antibacterial properties against Porphyromonas gingivalis, Prevotella intermedia, and Solobacterium moorei, with minimal inhibitory concentrations ranging from 2 to 80 µg/ml. These compounds have the potential to mitigate bacterial volatile sulfur compound (VSC) production, making them suitable for managing halitosis. Furthermore, they hold promise for novel approaches to modulate hosts′ responses in conditions involving cytokines and MMPs, such as periodontitis. Licoricidin may find application in topically applied anti-aging formulations by preventing UVA-induced photoaging through ROS scavenging, ultimately limiting MMP-1 activity.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service