Skip to Content
Merck
All Photos(1)

Key Documents

SMB01042

Sigma-Aldrich

Allocryptopin

≥90% (LC/MS-ELSD)

Synonym(s):

β-Allocryptopine, γ-Homochelidonine, Fagarine I

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€306.00

€306.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€306.00

About This Item

Empirical Formula (Hill Notation):
C21H23NO5
CAS Number:
Molecular Weight:
369.41
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

€306.00


Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

369.41

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

N1(CCc2c(cc4c(c2)OCO4)C(=O)Cc3c(c(c(cc3)OC)OC)C1)C

InChI

1S/C21H23NO5/c1-22-7-6-14-9-19-20(27-12-26-19)10-15(14)17(23)8-13-4-5-18(24-2)21(25-3)16(13)11-22/h4-5,9-10H,6-8,11-12H2,1-3H3

InChI key

HYBRYAPKQCZIAE-UHFFFAOYSA-N

General description

Allocryptopin, an isoquinoline alkaliod, is a natural product commonly available from Allocryptopinus japonicus, Glaucium arabicum, Zanthoxylum beecheyanum, Berberis integerrima, and Macleaya cordata. This secondary metabolite derived from plant sources exhibits various biological activities such as anti-inflammatory, antiviral, and antitumor properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Allocryptopine has been reported to possess strong antibacterial, anti-inflammatoty, antiparasitic, antineoplastic and anti-addictive activities. Allocryptopine inhibits human acetylcholinesterase and butyrylcholinesterase. It blocks hERG current in HEK293 cells. Allocryptopine strongly enhanced the I(peak) of the T353I channel by enhancing the plasma membrane (PM) expression of Nav1.5 and rescued defective trafficking after co-incubation with HEK293 cells that carry mutation channel 24 h.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service