Skip to Content
Merck
All Photos(1)

Key Documents

SMB01016

Sigma-Aldrich

Pomolic acid

≥90% (LC/MS-ELSD)

Synonym(s):

19α-Hydroxyursolic acid, Benthamic acid, Randialic acid A

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€418.00

€418.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€418.00

About This Item

Empirical Formula (Hill Notation):
C30H48O4
CAS Number:
Molecular Weight:
472.70
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

€418.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

472.7

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O[C@]1([C@@H]2[C@@](CC[C@]3([C@]4([C@@H]([C@@]5([C@H](C([C@H](CC5)O)(C)C)CC4)C)CC=C32)C)C)(CC[C@H]1C)C(=O)O)C

InChI

1S/C30H48O4/c1-18-10-15-30(24(32)33)17-16-27(5)19(23(30)29(18,7)34)8-9-21-26(4)13-12-22(31)25(2,3)20(26)11-14-28(21,27)6/h8,18,20-23,31,34H,9-17H2,1-7H3,(H,32,33)/t18-,20+,21-,22+,23-,26+,27-,28-,29-,30+/m1/s1

InChI key

ZZTYPLSBNNGEIS-OPAXANQDSA-N

General description

Natural product derived from plant source.
Pomolic acid, a pentacyclic triterpene, is a bioactive natural compound commonly derived from plants such as Euscaphis japonica, Centella asiatica, Picramnia sellowii, and Cecropia pachystachya. Current research suggests that this metabolite acts as an inhibitor and may exhibit diverse biological activities, including anticancer, anti-inflammatory, antiviral, apoptotic, and antihypertensive properties.

Application

Pomolic acid is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Pomolic acid is highly effective in inhibiting cell growth and inducing apoptosis.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phuong Thien Thuong et al.
Biological & pharmaceutical bulletin, 29(4), 830-833 (2006-04-06)
A new ursane-type triterpenoid, weigelic acid (1), and seven known compounds, ursolic acid (2), ilekudinol A (3), corosolic acid (4), ilekudinol B (5), esculentic acid (6), pomolic acid (7), and asiatic acid (8) were isolated from the leaf and stem
Guillermo Schinella et al.
Planta medica, 74(3), 215-220 (2008-02-09)
The dichloromethane extract and pomolic acid ( 5) obtained from leaves of Cecropia pachystachya both reduced carrageenan-induced paw oedema in mice. Interestingly, while the triterpenoid inhibited the in vivo production of interleukin-1beta by 39 %, it had no effect on
Y Kashiwada et al.
Journal of natural products, 61(9), 1090-1095 (1998-09-28)
Oleanolic acid (1) was identified as an anti-HIV principle from several plants, including Rosa woodsii (leaves), Prosopis glandulosa (leaves and twigs), Phoradendron juniperinum (whole plant), Syzygium claviflorum (leaves), Hyptis capitata (whole plant), and Ternstromia gymnanthera (aerial part). It inhibited HIV-1
Studies on constituents of medicinal plants. XIII. Constituents of the pericarps of the capsules of Euscaphis japonica Pax.
K Takahashi et al.
Chemical & pharmaceutical bulletin, 22(3), 650-653 (1974-03-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service