Skip to Content
Merck
All Photos(1)

Key Documents

SMB01012

Sigma-Aldrich

Mussaenosidic acid

≥90% (LC/MS-ELSD)

Synonym(s):

Cyclopenta[c]pyran-4-carboxylic acid, 1-(β-D-glucopyranosyloxy)-1,4a,5,6,7,7a-hexahydro-7-hydroxy-7-methyl-, [1S-(1α,4aα,7α,7aα)]- (ZCI)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€384.00

€384.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€384.00

About This Item

Empirical Formula (Hill Notation):
C16H24O10
CAS Number:
Molecular Weight:
376.36
UNSPSC Code:
12352205
NACRES:
NA.25

€384.00


Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

376.36

solubility

water: soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]2OC=C([C@@H]3[C@H]2[C@](CC3)(O)C)C(=O)O

InChI key

VLCHQFXSBHIBRV-NJPMDSMTSA-N

General description

Mussaenosidic acid, an iridoid glucoside, is a bioactive natural product commonly available from Melampyrum Sp., Pedicularis kerneri, Lagochilus ilicifolius, Vitex negundo, Barleria lupulina and Kickxia Sp. Plants.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and biochemical research.

Biochem/physiol Actions

Mussaenosidic acid shows weak antiglycation activity.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adnan Amin et al.
Frontiers in pharmacology, 8, 232-232 (2017-05-17)
Background and Aims:Kickxia ramosissima (Wall.) Janch (or Nanorrhinum ramosissimum (Wall.) Betsche is a well-known medicinal plant in Pakistan that is traditionally used in diabetic and inflammatory conditions. Because little information is available on its phytochemical composition, a range of constituents

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service