Skip to Content
Merck
All Photos(1)

Key Documents

SMB00445

Sigma-Aldrich

Ganoderic acid A

≥98% (HPLC)

Synonym(s):

(2R,6R)-6-[(5R,7S,10S,13R,14R,15S,17R)-7,15-dihydroxy-4,4,10,13,14-pentamethyl-3,11-dioxo-2,5,6,7,12,15,16,17-octahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-methyl-4-oxoheptanoic acid, (7β,15α,25R)-7,15-Dihydroxy-3,11,23-trioxo-lanost-8-en-26-oic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€386.00

€386.00


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€386.00

About This Item

Empirical Formula (Hill Notation):
C30H44O7
CAS Number:
Molecular Weight:
516.67
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

€386.00


Estimated to ship on29 March 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC(C(C)CC(C[C@@H](C)[C@H]1C[C@H](O)[C@@]([C@]1(C)CC2=O)(C)C3=C2[C@]4(C)C(C[C@@H]3O)C(C)(C)C(CC4)=O)=O)=O

InChI

1S/C30H44O7/c1-15(10-17(31)11-16(2)26(36)37)18-12-23(35)30(7)25-19(32)13-21-27(3,4)22(34)8-9-28(21,5)24(25)20(33)14-29(18,30)6/h15-16,18-19,21,23,32,35H,8-14H2,1-7H3,(H,36,37)/t15-,16?,18-,19+,21?,23+,28+,29-,30+/m1/s1

InChI key

DYOKDAQBNHPJFD-ZQEHRSJRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ganoderic Acid A or GAA is one of the most abundant triterpenoids that is found in Ganoderma lucidum fungi.[1] The chemical structure of GAA has a tetracyclic ring with a double bond and terminal carboxyl group on the branch.[1]

Application

Ganoderic acid A has been used:
  • to study its protective effects on hypoxia-induced rat cardiomyocytes (H9c2) cell injury[2]
  • as a reference standard to study its inhibitory and antiviral effects against groundnut bud necrosis virus (GBNV) infection in cowpea plants[3]
  • as a standard in Fourier transformed-infrared (FT-IR) spectroscopy for the analysis of secondary metabolites from Ganoderma lucidum[4]

Biochem/physiol Actions

Ganoderic Acid A (GAA) improves lipid metabolism, gut microbiota composition, and hyperlipidemia in high-fat-diet (HFD)-fed mice.[1] It is proven to be a potential therapeutic anticancer agent in several in vitro studies by suppressing cell proliferation in different cancer cells such as breast cancer, human hepatocellular carcinoma cells, and osteosarcoma.[5] GAA displays neuroprotective activities against depression-like behaviors, inflammation, and neuronal damage in the post-stroke depression (PSD) rat model.[6] It also displays anti-human immunodeficiency virus (HIV) activity.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Wang et al.
Molecular medicine reports, 16(4), 3894-3900 (2017-07-22)
Ganoderic acid A (GA‑A), a triterpenoid, has been demonstrated to suppress cell proliferation in various cancers, including breast cancer and osteosarcoma. However, its effect on human hepatocellular carcinoma (HCC) remains to be elucidated. The present study aimed to investigate the
Yanhua Chang et al.
Phytotherapy research : PTR, 33(5), 1448-1456 (2019-03-07)
Effects of ganoderic acid A (GAA), a lanostane triterpene, on hypoxia-ischemia encephalopathy (HIE) remain unclear. We aimed to figure out the specific role of GAA in hypoxia-treated neural stem cells (NSCs) as well as the regulatory mechanisms. Primary rat NSCs
Jiahua Jiang et al.
International journal of molecular medicine, 21(5), 577-584 (2008-04-22)
Structurally related lanostane-type triterpenes, ganoderic acid A, F and H (GA-A, GA-F, GA-H), were identified in an oriental medicinal mushroom Ganoderma lucidum. In the present study we evaluated the effect of GA-A, GA-H and GA-F on highly invasive human breast
Xiaohong Zhang et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 50(6), 2086-2096 (2018-11-12)
Ganoderic acid A (GAA) isolated from Ganoderma lucidum, shows various benefit activities, such as anti-tumor activity, anti-HIV activity and hepatoprotective activity. However, the potential effects of GAA on hypoxia-induced injury of cardiomyocytes are still unclear. In this study, we aimed
Bingji Ma et al.
North American journal of medical sciences, 3(11), 495-498 (2012-03-01)
Recently a series of triterpenoids were isolated from ganoderma spores and have drawn the attention of chemists and pharmacists. The aim of this review is to summarize the triterpenoids and their bioactivities of ganoderma spores. The chemical and biological literatures

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service