Skip to Content
Merck
All Photos(1)

Key Documents

SMB00379

Sigma-Aldrich

Schisantherin A

≥98% (HPLC)

Synonym(s):

Gomisin C, Schizantherin A, Wuweizi ester A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H32O9
CAS Number:
Molecular Weight:
536.57
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC1(O)C(C)CC2=CC(OCO3)=C3C(OC)=C2C4=C(C=C(OC)C(OC)=C4OC)C1OC(C5=CC=CC=C5)=O

InChI

1S/C30H32O9/c1-16-12-18-13-21-25(38-15-37-21)26(35-5)22(18)23-19(14-20(33-3)24(34-4)27(23)36-6)28(30(16,2)32)39-29(31)17-10-8-7-9-11-17/h7-11,13-14,16,28,32H,12,15H2,1-6H3

InChI key

UFCGDBKFOKKVAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Schisantherin A is a lignan isolated from the fruits of Schisandra chinesi.[1]
Schisantherin A is a mechanism-based inhibitor that competitively inhibits and irreversibly inactivates CYP3A4.[2]
Schisantherin A (StA) has neuroprotective activity and could be potential candidate for the treatment of neuron loss in neurodegenerative diseases including Parkinson′s disease.[3] It inhibits tumor progression and could be a potential drug target for cancer treatment.[4] StA blocks ATP-binding cassette transporters and restores the levels of anti-cancer drugs in cancer cells.[5] It also has anti-inflammatory property.[6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Schisantherin A suppresses interleukin-1β-induced inflammation in human chondrocytes via inhibition of NF-κB and MAPKs activation.
Liao S, et al.
European Journal of Pharmacology, 780, 65-70 (2016)
Synthesis of the first unnatural schisantherins and their effects in multidrug-resistant cancer cells.
Schobert R, et al.
Tetrahedron Letters, 49(21), 3359-3362 (2008)
Lignans from Schisandra sphenathera Rehd. et Wils. and semisynthetic schisantherin A analogues: absolute configuration, and their estrogenic and anti-proliferative activity.
Liu H W, et al.
European Journal of Medicinal Chemistry, 59, 265-273 (2013)
Discovery of novel anti-parkinsonian effect of schisantherin A in in vitro and in vivo.
Sa F, et al.
Neuroscience Letters, 593, 7-12 (2015)
Schisandra Chinensis-derived antioxidant activities in `Sheng Mai San?a compound formulation, in vivo and in vitro.
Ko K M, et al.
Phytotherapy Research, 9(3), 203-206 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service