Skip to Content
Merck
All Photos(1)

Key Documents

SMB00354

Sigma-Aldrich

Fraxinellone

≥98% (HPLC)

Synonym(s):

3-(3-Furyl)-3a,4,5,6-tetrahydro-3a,7-dimethylphthalide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16O3
CAS Number:
Molecular Weight:
232.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC1=C(C(O[C@H]2C3=COC=C3)=O)[C@@]2(C)CCC1

InChI

1S/C14H16O3/c1-9-4-3-6-14(2)11(9)13(15)17-12(14)10-5-7-16-8-10/h5,7-8,12H,3-4,6H2,1-2H3/t12-,14+/m0/s1

InChI key

XYYAFLHHHZVPRN-GXTWGEPZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fraxinellone is a terpenoid found in the root bark of Dictamnus albus. It belongs to a small group of degraded limonoids.[1][2]

Biochem/physiol Actions

Fraxinellone possesses neuroprotective and vasorelaxing activities as well as anti-inflammatory properties.[3] Fraxinellone has been used as a potent therapeutic for T-cell-mediated liver disorders in humans.[1]

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective triggering of apoptosis of concanavalin A-activated T cells by fraxinellone for the treatment of T-cell-dependent hepatitis in mice
Sun Y, et al.
Biochemical Pharmacology, 77(11), 1717-1724 (2009)
Fraxinellone inhibits lipopolysaccharide-induced inducible nitric oxide synthase and cyclooxygenase-2 expression by negatively regulating nuclear factor-kappa B in RAW 264.7 macrophages cells.
Kim J H, et al.
Biological & Pharmaceutical Bulletin, 32(6), 1062-1068 (2009)
Antimutagenic Activity of Isofraxinellone from Dictamnus dasycarpus
Miyazawa M, et al.
Journal of Agricultural and Food Chemistry, 43(6), 1428-1431 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service