Skip to Content
Merck
All Photos(1)

Documents

SMB00252

Sigma-Aldrich

Osmanthuside H

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28O11
CAS Number:
Molecular Weight:
432.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC[C@@]1(O)CO[C@@H](OC[C@H]2O[C@@H](OCCc3ccc(O)cc3)[C@H](O)[C@@H](O)[C@@H]2O)[C@@H]1O

InChI

1S/C19H28O11/c20-8-19(26)9-29-18(16(19)25)28-7-12-13(22)14(23)15(24)17(30-12)27-6-5-10-1-3-11(21)4-2-10/h1-4,12-18,20-26H,5-9H2/t12-,13-,14+,15-,16+,17-,18-,19-/m1/s1

InChI key

IVRQZYXJBVMHCW-OTCFHACESA-N

General description

Natural product derived from plant source.
Osmanthuside H is a chemical constituent, isolated from stem barks of Fraxinus paxiana.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phenolic compounds and their anti-oxidative properties and protein kinase inhibition from the Chinese mangrove plant Laguncularia racemosa.
Cui S, et al.
Phytochemistry, 71(4), 435-442 (2010)
M Sugiyama et al.
Phytochemistry, 32(6), 1553-1555 (1993-04-01)
Three new phenylethanoid glycosides, 2-(4-hydroxyphenyl)ethyl-beta-D-apiosyl-(1----6)-beta-D-glucopy ranoside(osmanthuside H), 2-(4-hydroxyphenyl)ethyl 5-O-trans-p-coumaroyl-beta-D-apiosyl(1----6)-beta-D-gluc+ ++ opy ranoside (osmanthuside I) and 2-(4-hydroxyphenyl)ethyl 5-O-trans-feruloyl-beta-D-apiosyl-(1----6)-beta-D-glucopy r ano side (osmanthuside J), were isolated from the bark of Osmanthus asiaticus. The structures were elucidated on the basis of spectroscopic evidence.
Zhi-jing Ma et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 33(16), 1990-1993 (2008-12-18)
To investigate the chemical constituents of Fraxinus paxiana. The chemical constituents were isolated and purified by chromatographic techniques and the structures of the compounds were identified with or by spectroscopic methods. Fifteen compounds were obtained from the methanol extract of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service