Skip to Content
Merck
All Photos(1)

Key Documents

SMB00247

Sigma-Aldrich

1,3,6-Trigalloylglucose

≥95% (LC/MS-ELSD)

Synonym(s):

1,3,6-Tri-O-galloyl-β-D-glucopyranose, 1,3,6-Tri-O-galloyl-β-D-glucose, Tannic acid (Corilagin)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€461.00

€461.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
€461.00

About This Item

Empirical Formula (Hill Notation):
C27H24O18
CAS Number:
Molecular Weight:
636.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

€461.00


Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (LC/MS-ELSD)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O=C(C1=CC(O)=C(O)C(O)=C1)O[C@H]2[C@H](O)[C@@H](OC(C3=CC(O)=C(O)C(O)=C3)=O)[C@H](O)[C@@H](COC(C4=CC(O)=C(O)C(O)=C4)=O)O2

InChI

1S/C27H24O18/c28-11-1-8(2-12(29)18(11)34)24(39)42-7-17-21(37)23(44-25(40)9-3-13(30)19(35)14(31)4-9)22(38)27(43-17)45-26(41)10-5-15(32)20(36)16(33)6-10/h1-6,17,21-23,27-38H,7H2/t17-,21-,22-,23+,27+/m1/s1

InChI key

RNKMOGIPOMVCHO-SJMVAQJGSA-N

General description

Natural product derived from plant source.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Danh C Vu et al.
Journal of agricultural and food chemistry, 66(17), 4503-4511 (2018-04-18)
Black walnuts ( Juglans nigra L.) are highly valued for producing phenolic-enriched nuts. The objectives of this study were to identify and characterize the phenolic contents of 11 different black walnut cultivars and compare the levels of these phenolics between
Alessia Panusa et al.
Phytochemistry, 115, 79-88 (2015-02-24)
The aim of this study was to get a rapid metabolic fingerprinting and to gain insight into the metabolic profiling of Arctostaphylos pungens H. B. K., a plant morphologically similar to Arctostaphylos uva-ursi (L.) Spreng. (bearberry) but with a lower

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service