Skip to Content
Merck
All Photos(1)

Key Documents

SMB00235

Sigma-Aldrich

Evodol

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€461.00

€461.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
€461.00

About This Item

Empirical Formula (Hill Notation):
C26H28O9
CAS Number:
Molecular Weight:
484.50
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

€461.00


Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC1(C)O[C@H]2CC(=O)OC[C@]23[C@H]4CC[C@@]5(C)C(OC(=O)[C@H]6O[C@@]56[C@]4(C)C(=O)C(O)=C13)c7ccoc7

InChI

1S/C26H28O9/c1-22(2)17-16(28)18(29)24(4)13(25(17)11-32-15(27)9-14(25)34-22)5-7-23(3)19(12-6-8-31-10-12)33-21(30)20-26(23,24)35-20/h6,8,10,13-14,19-20,28H,5,7,9,11H2,1-4H3/t13-,14-,19?,20+,23-,24-,25-,26+/m0/s1

InChI key

SNGHLUWTFLYPMT-FBIYPHEJSA-N

General description

Natural product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service