Skip to Content
Merck
All Photos(1)

Key Documents

SMB00181

Sigma-Aldrich

Liriodendrin

≥85% (LC/MS-ELSD)

Synonym(s):

(+)-Syringaresinol di-O-β-glucopyranoside, Acanthoside D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H46O18
CAS Number:
Molecular Weight:
742.72
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥85% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1cc(cc(OC)c1O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H]3OCC4C3CO[C@@H]4c5cc(OC)c(OC6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)c(OC)c5

InChI

1S/C34H46O18/c1-43-17-5-13(6-18(44-2)31(17)51-33-27(41)25(39)23(37)21(9-35)49-33)29-15-11-48-30(16(15)12-47-29)14-7-19(45-3)32(20(8-14)46-4)52-34-28(42)26(40)24(38)22(10-36)50-34/h5-8,15-16,21-30,33-42H,9-12H2,1-4H3/t15-,16-,21+,22+,23+,24+,25-,26-,27+,28+,29+,30+,33-,34?/m0/s1

InChI key

FFDULTAFAQRACT-MEAGMGCTSA-N

General description

Natural product derived from plant source.

Biochem/physiol Actions

Liriodendrin has shown anti-inflammatory and antinociceptive properties, the administration of liriodendrin caused a reduction in acute paw edema induced by carrageenan in rats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chu Van Men et al.
Archives of pharmacal research, 34(12), 2065-2071 (2012-01-03)
A quantitative and pattern recognition analyses were conducted for quality evaluation of Kalopanacis Cortex (KC) using HPLC. For quantitative analysis, four bioactive compounds, liriodendrin, pinoresinol O-β-D-glucopyranoside, acanthoside B and kalopanaxin B, were determined. The analysis method was optimized and validated
Jian-fei Chao et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 31(13), 1078-1080 (2006-10-20)
To investigate the chemical constituents from the branch of Broussonetia papyrifera. Column chromatographic methods were used to isolate the chemical constituents. ESI-MS and NMR methods were employed for their structural elucidation. Six compounds were isolated and identified as (2S)-7, 3'-dihydroxy-4'-methoxyflavan
Hyun-Ju Jung et al.
Planta medica, 69(7), 610-616 (2003-08-05)
In the present study, liriodendrin isolated by activity-guided fractionation from the ethyl acetate (EtOAc) extracts of the stem bark of Acanthopanax senticosus, was evaluated for anti-inflammatory and antinociceptive activities. Liriodendrin (5, 10 mg/kg/day, p. o.) significantly inhibited the increase of
Tomoko Sano et al.
Journal of natural medicines, 64(3), 257-265 (2010-03-11)
Boi and its original plant Sinomenium acutum from Japan were compared with Seifuto and its botanical origins from China in terms of their internal transcribed spacer (ITS) sequences and major chemical components. Boi, Seifuto, and their botanical origins overall showed
L B Kardono et al.
Journal of natural products, 53(6), 1447-1455 (1990-11-01)
By bioactivity-directed fractionation, six cytotoxic constituents have been characterized from the bark of Plumeria rubra collected in Indonesia. Three iridoids, fulvoplumierin [1], allamcin [2], and allamandin [3], as well as 2,5-dimethoxy-p-benzoquinone [4], were found to be active constituents of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service