Skip to Content
Merck
All Photos(4)

Key Documents

SHH0002

Sigma-Aldrich

Novec 7100 Engineered Fluid

greener alternative

≥99.5% purity, liquid

Synonym(s):

methoxy-nonafluorobutane; HFE-7100

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€191.00
1 L
€323.00

€191.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
€191.00
1 L
€323.00

About This Item

Empirical Formula (Hill Notation):
C5H3OF9
Molecular Weight:
250.06
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€191.00


Check Cart for Availability

Request a Bulk Order

Product Name

Novec 7100 Engineered Fluid, ≥99.5%

Assay

≥99.5%

form

liquid

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

titration: suitable

color

colorless to yellow

mp

-135 °C

application(s)

diagnostic assay manufacturing
hematology
histology

greener alternative category

storage temp.

room temp

SMILES string

FC(C(F)(F)C(F)(F)OC)(F)C(F)(F)F.FC(C(F)(F)OC)(C(F)(F)F)C(F)(F)F

InChI

1S/2C5H3F9O/c1-15-5(13,14)3(8,9)2(6,7)4(10,11)12;1-15-5(13,14)2(6,3(7,8)9)4(10,11)12/h2*1H3

InChI key

SXYHZEQKWNODPB-UHFFFAOYSA-N

General description

Novec 7100 fluid consists of two inseparable isomers with essentially identical properties. These are (CF3)2CFCF2OCH3 (CAS No. 163702-08-7) and CF3CF2CF2CF2OCH3 (CAS No. 163702-07-6).
Novec 7100 engineered fluid is a proprietary methoxy-nonafluorobutane from 3M. This hydrofluoroether is a clear, colorless and low-odor fluid intended to replace ozone-depleting substances (ODSs) and compounds with high global warming potential (GWP) in many applications. It has zero ozone depletion potential. High boiling point and low surface tension properties make it ideal for use in vapor degreasing applications as a neat (pure), azeotropic component or co-solvent parts cleaner. In addition, its chemical and thermal stability, non-flammability and low toxicity make it useful for many other industrial and specialty solvent applications.

Application

  • Cleaning and rinsing agent
-Heavy-duty cleaning (co-solvent) – heavy oils, greases, fluxes
-Medium-duty cleaning (azeotrope)– oils, greases, waxes
-Light-duty cleaning (neat)– particulates, fluorolubes, light oils, fluoropolymer
  • Lubricant carrier: fluorocarbons, hydrocarbons, and silicones
  • Spot-free water drying agent (with surfactants added)
  • Specialty solvents, dispersion media, reaction media
  • Spray contact cleaner
  • CFC, HCFC, HFC and PFC replacement
  • Dielectric test media
  • Heat transfer
  • Preservation of biological specimens
  • Cryofreezing solution
  • Non-flammable
  • Non-conductive
  • Compatible with plastics
  • Low toxicity
  • Low Global Warming Potential (GWP)
  • Zero ozone depletion potential
  • VOC-exempt solvent (US EPA)
  • Low surface tension penetrates tight spaces for improved cleaning

Packaging

Packaged and distributed by Sigma-Aldrich

Legal Information

3M is a trademark of 3M Company
Novec is a trademark of 3M Company

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malte Plidschun et al.
Light, science & applications, 10(1), 57-57 (2021-03-17)
Strong focusing on diffraction-limited spots is essential for many photonic applications and is particularly relevant for optical trapping; however, all currently used approaches fail to simultaneously provide flexible transportation of light, straightforward implementation, compatibility with waveguide circuitry, and strong focusing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service