Skip to Content
Merck
All Photos(1)

Key Documents

SCP0234

Sigma-Aldrich

PTH Peptide

≥95% (HPLC), lyophilized powder, bovine, 3-34

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€320.00
1 MG
€548.00

€320.00


Usually ships in 2 weeks.


Select a Size

Change View
500 μG
€320.00
1 MG
€548.00

About This Item

Empirical Formula (Hill Notation):
C177H279N53O48
Molecular Weight:
3917.44
UNSPSC Code:
12352209
NACRES:
NA.32

€320.00


Usually ships in 2 weeks.

Product Name

[Nle8,18,Tyr34]-PTH 3-34 bovine,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥85%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ser-Glu-Ile-Gln-Phe-Nle-His-Asn-Leu-Gly-Lys-His-Leu-Ser-Ser-Nle-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Tyr-NH2

Application

Parathyroid hormone (PTH) is an 84 amino acid hormone involved in calcium regulation in counteraction to calcitonin. PTH regulates serum phosphate levels and vitamin D biosynthesis. N-terminal peptides of PTH are used to elicit and study a wide variety of responses in target cells. PTH N-terminal peptides include: PTH (1-34); [Nle8,18,Tyr34]-PTH 1-34; PTH (2-34); [Nle8,18,Tyr34]-PTH (2-34); PTH (3-34); [Nle8,18,Tyr34]-PTH (3-34); PTH (4-34) and PTH (5-34).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Estelle N Bianchi et al.
Bone, 45(4), 716-725 (2009-06-30)
Interaction of the cytoplasmic adaptor molecule beta-arrestin2 with the activated parathyroid hormone (PTH)/PTHrP receptor inhibits G protein mediated signaling and triggers MAPKs signaling. In turn, the effects of both intermittent (i.) and continuous (c.) PTH on bone are altered in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service